missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
4-Anilino-4-oxobutanoic Acid-d5, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-Anilino-d5-4-oxobutanoic Acid |
| CAS | 840529-98-8 |
| Molekylformel | C10 2H5 H6 N O3 |
| Kvantitet | 10 mg |
| Synonym | Butanoic acid, 4-oxo-4-(phenyl-2,3,4,5,6-d5-amino)-, Butanoic acid, 4-oxo-4-(phenyl-d5-amino)- (9CI), 4-Oxo-4-(phenyl-2,3,4,5,6-d5-amino)butanoic acid, Succinanilic acid-d5, 4-Oxo-4-(phenylamino)butanoic acid D5 (phenyl-D5), 4-(Anilino-d5)-4-oxobutanoic acid, 4-Oxo-4-(N-phenyl-d5-amino)butanoic acid, N-(Phenyl-d5)butanedioic acid monoamide, N-(Phenyl-d5)succinamic acid, Succinic acid monoanilide-d5, Succinic monoanilide-d5 |
| InChI formel | InChI=1S/C10H11NO3/c12-9(6-7-10(13)14)11-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,11,12)(H,13,14)/i1D,2D,3D,4D,5D |
| LEDER | [2H]c1c([2H])c([2H])c(NC(=O)CCC(=O)O)c([2H])c1[2H] |
| IUPAC-namn | 4-oxo-4-(2,3,4,5,6-pentadeuterioanilino)butanoic acid |
| Molekylvikt (g/mol) | 198.23 |
| Formel vikt | 198.1053 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?