missing translation for 'onlineSavingsMsg'
Learn More

4-Azaindole, 98%, Thermo Scientific™

Catalog Number p-7077629
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
272-49-1
C7H6N2
118.14
MFCD00971977
XWIYUCRMWCHYJR-UHFFFAOYSA-N
4-azaindole, 1h-pyrrolo 3,2-b pyridine, 1,4-diazaindene, pyrrolopyridine, 4h-pyrrolo 3,2-b pyridine, 1h-pyrrolo 3.2-b pyridine, zlchem 17, pubchem7675, acmc-1clkx
9226
1H-pyrrolo[3,2-b]pyridine
N1C=CC2=NC=CC=C12
This item is not returnable. View return policy
272-49-1
C7H6N2
118.14
MFCD00971977
XWIYUCRMWCHYJR-UHFFFAOYSA-N
4-azaindole, 1h-pyrrolo 3,2-b pyridine, 1,4-diazaindene, pyrrolopyridine, 4h-pyrrolo 3,2-b pyridine, 1h-pyrrolo 3.2-b pyridine, zlchem 17, pubchem7675, acmc-1clkx
9226
1H-pyrrolo[3,2-b]pyridine
N1C=CC2=NC=CC=C12
Melting Point 127.0°C to 130.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 118.14
Percent Purity 98%
Chemical Name or Material 4-Azaindole
compliance-icons
Product Identifier
  • 4-Azaindole, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'