Läs mer
4-Azido-L-phenylalanine Hydrochloride, TRC
Produktkod. 17912945
Change view
Kvantitet:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17912945 500 mg
17904495 25 mg
17971635 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17912945 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA922600500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34670-43-4
Molekylformel C9 H10 N4 O2 . Cl H
Molekylvikt (g/mol) 242.662
Synonym 3-(p-Azidophenyl)-L-alanine Hydrochloride, 4-Azidophenylalanine Hydrochloride, p-Azido-L-phenylalanine Hydrochloride, p-Azidophenylalanine Hydrochloride
IUPAC-namn (2S)-2-amino-3-(4-azidophenyl)propanoic acid;hydrochloride
LEDER Cl.N[C@@H](Cc1ccc(cc1)N=[N+]=[N-])C(=O)O
Specifikationer
Kemiskt namn eller material 4-Azido-L-phenylalanine Hydrochloride
InChI formel InChI=1S/C9H10N4O2.ClH/c10-8(9(14)15)5-6-1-3-7(4-2-6)12-13-11;/h1-4,8H,5,10H2,(H,14,15);1H/t8-;/m0./s1
Kvantitet 500 mg
Formel vikt 242.0571