Läs mer
4-Benzhydryl-1-piperazineethanol Dihydrochloride, TRC
Produktkod. 30343271
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30343271 1 g
1 options
Produktkod. 30343271 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB1969551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(4-Benzhydrylpiperazin-1-yl)ethan-1-ol Dihydrochloride
CAS 108983-83-1
Molekylformel C19 H24 N2 O . 2 Cl H
Kvantitet 1 g
Synonym 4-(Diphenylmethyl)-1-piperazineethanol Hydrochloride (1:2), 1-Piperazineethanol, 4-diphenylmethyl-, dihydrochloride (6CI), 4-Benzhydryl-1-piperazineethanol Dihydrochloride, 2-(4-(Diphenylmethyl)piperazin-1-yl)ethanol Dihydrochloride, Deschlorodesacetic Cetirizine
InChI formel InChI=1S/C19H24N2O.2ClH/c22-16-15-20-11-13-21(14-12-20)19(17-7-3-1-4-8-17)18-9-5-2-6-10-18;;/h1-10,19,22H,11-16H2;2*1H
LEDER Cl.Cl.OCCN1CCN(CC1)C(c2ccccc2)c3ccccc3
IUPAC-namn 2-(4-benzhydrylpiperazin-1-yl)ethanol;dihydrochloride
Molekylvikt (g/mol) 369.33
Formel vikt 368.1422