Learn More
4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione, TRC
Product Code. 30329633
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity Unit Size
30329633 1 mg 1mg
1 options
Product Code. 30329633 Supplier Toronto Research Chemicals Supplier No. TRCB2857251MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
CAS 327036-89-5
Molecular Formula C10 H10 N2 O2 S
Quantity 1 mg
Synonym TDZD 8 - Bio-X TM
InChI Formula InChI=1S/C10H10N2O2S/c1-11-9(13)12(10(14)15-11)7-8-5-3-2-4-6-8/h2-6H,7H2,1H3
SMILES CN1SC(=O)N(Cc2ccccc2)C1=O
IUPAC Name 4-benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
Molecular Weight (g/mol) 222.26
Formula Weight 222.0463