Läs mer
4-Benzyloxy-1-bromobutane (~90%), TRC
Produktkod. 30347055
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30347055 100 mg
1 options
Produktkod. 30347055 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB291445100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Benzyloxy-1-bromobutane
CAS 60789-54-0
Molekylformel C11H15BrO
Kvantitet 100 mg
Synonym Ether, benzyl 4-bromobutyl (6CI), [(4-Bromobutoxy)methyl]benzene, 1-(Benzyloxy)-4-bromobutane, 1-Bromo-4-(benzyloxy)butane, 4-(Benzyloxy)butyl bromide, 4-Bromobutyl phenylmethyl ether, Benzyl 4-bromobutyl ether
InChI formel InChI=1S/C11H15BrO/c12-8-4-5-9-13-10-11-6-2-1-3-7-11/h1-3,6-7H,4-5,8-10H2
LEDER BrCCCCOCc1ccccc1
IUPAC-namn 4-bromobutoxymethylbenzene
Molekylvikt (g/mol) 243.14
Formel vikt 242.0306