Läs mer
4-(Benzyloxy)-2-butynoic Acid Methyl Ester, TRC
Produktkod. 30334420
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30334420 100 mg
1 options
Produktkod. 30334420 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB258800100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(Benzyloxy)-2-butynoic Acid Methyl Ester
CAS 149125-61-1
Molekylformel C12H12O3
Kvantitet 100 mg
Synonym 4-(Phenylmethoxy)-2-butynoic Acid Methyl Ester, Methyl 4-Benzyloxy-2-butynoate
InChI formel InChI=1S/C12H12O3/c1-14-12(13)8-5-9-15-10-11-6-3-2-4-7-11/h2-4,6-7H,9-10H2,1H3
LEDER O=C(OC)C#CCOCC1=CC=CC=C1
IUPAC-namn methyl 4-(benzyloxy)but-2-ynoate
Molekylvikt (g/mol) 204.22
Formel vikt 204.08