Läs mer
4-Benzyloxy-2-chloro-5-hydroxypyrimidine, 95%, Thermo Scientific™
Produktkod. 15423587
Kvantitet:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
Produktkod. 15423587
Brand: Thermo Scientific Alfa Aesar H26993.MD
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 885952-28-3
Molekylformel C11H9ClN2O2
Molekylvikt (g/mol) 236.66
MDL-nummer MFCD06798283
InChI-nyckel URJRKXXIIWYWIS-UHFFFAOYSA-N
Synonym 4-benzyloxy-2-chloropyrimidin-5-ol, 4-benzyloxy-2-chloro-5-hydroxypyrimidine, 4-benzyloxy-2-chloro-5-pyrimidinol, 5-pyrimidinol,2-chloro-4-phenylmethoxy
PubChem CID 42552982
IUPAC-namn 2-chloro-4-phenylmethoxypyrimidin-5-ol
LEDER OC1=CN=C(Cl)N=C1OCC1=CC=CC=C1
Specifikationer
Kemiskt namn eller material 4-Benzyloxy-2-chloro-5-hydroxypyrimidine
Kvantitet 250 mg
Formel vikt 236.66
Procent renhet 95%