Läs mer
4-Benzyloxy-3-methoxybenzoic Acid Methyl Ester, TRC
Produktkod. 30330694
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30330694 500 mg
1 options
Produktkod. 30330694 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB286455500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Benzyloxy-3-methoxybenzoic Acid Methyl Ester
CAS 56441-97-5
Molekylformel C16H16O4
Kvantitet 500 mg
Synonym Methyl 3-Methoxy-4-(benzyloxy)benzoate, Methyl 4-(Benzyloxy)-3-methoxybenzoate
InChI formel InChI=1S/C16H16O4/c1-18-15-10-13(16(17)19-2)8-9-14(15)20-11-12-6-4-3-5-7-12/h3-10H,11H2,1-2H3
LEDER COC(=O)c1ccc(OCc2ccccc2)c(OC)c1
IUPAC-namn methyl 3-methoxy-4-phenylmethoxybenzoate
Molekylvikt (g/mol) 272.3
Formel vikt 272.1049