Läs mer
4-Benzyloxyphenylacetonitrile, 97%, Thermo Scientific™
Produktkod. 11362257
Change view
Kvantitet:
250 mg
1 g
5 g
Förpackningsstorlek:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11362257 250 mg 250mg
11342257 1 g 1g
11352257 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 11362257 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer L09771.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 838-96-0
Molekylformel C15H13NO
Molekylvikt (g/mol) 223.28
MDL-nummer MFCD00016400
InChI-nyckel QKEYZRVDFZDOEP-UHFFFAOYSA-N
Synonym 4-benzyloxyphenylacetonitrile, 2-4-benzyloxy phenyl acetonitrile, 4-benzyloxybenzyl cyanide, 4-benzyloxy phenyl acetonitrile, 2-4-phenylmethoxy phenyl ethanenitrile, acmc-20amoi, pubchem15926, 4-benzyloxybenzeneacetonitrile, p-benzyloxyphenyl acetonitrile
PubChem CID 522804
IUPAC-namn 2-(4-phenylmethoxyphenyl)acetonitrile
LEDER N#CCC1=CC=C(OCC2=CC=CC=C2)C=C1
Specifikationer
Kemiskt namn eller material 4-Benzyloxyphenylacetonitrile
Smältpunkt 66°C to 70°C
Kokpunkt 170°C to 173°C (0.1 mmHg)
Kvantitet 250 mg
Beilstein 2377584
Formel vikt 223.28
Procent renhet 97%