missing translation for 'onlineSavingsMsg'
Läs mer

4-(Bis(4-Hydroxyphenyl)methylene)cyclohexa-2,5-dienone, TRC

Produktkod. 30003276
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30003276 10 g
30136560 1 g
30138629 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003276 Supplier Toronto Research Chemicals Supplier No. TRCB43415010G

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-45-2
Molekylformel C19 H14 O3
Molekylvikt (g/mol) 290.31
Synonym Aurin, 4,4'-Dihydroxyfuchsone, Rosalic Acid, Pararosolic Acid, Spirit Aurine, Corallin, Aurin 555, C.I. 43800, 4,4'-Dihydroxyfuchsone, Corallin Spirit Soluble;
IUPAC-namn 4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
LEDER Oc1ccc(cc1)C(=C2C=CC(=O)C=C2)c3ccc(O)cc3

Specifications

Kemiskt namn eller material 4-(Bis(4-Hydroxyphenyl)methylene)cyclohexa-2,5-dienone
Kvantitet 10 g
InChI formel InChI=1S/C19H14O3/c20-16-7-1-13(2-8-16)19(14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15/h1-12,20-21H
Formel vikt 290.0943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.