missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Boc-1-(5-bromo-2-pyridyl)piperazine, 97%, Thermo Scientific Chemicals
426.82 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Boc-1-(5-bromo-2-pyridyl)piperazine is used as active pharmaceutical ingredient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Boc-1-(5-bromo-2-pyridyl)piperazine is used as active pharmaceutical ingredient.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
4-Boc-1-(5-bromo-2-pyridyl)piperazine is used as active pharmaceutical ingredient.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 83°C to 87°C |
Color | White to Yellow |
Quantity | 1 g |
UN Number | UN2811 |
Solubility Information | Soluble in water. |
Formula Weight | 342.23 |
Percent Purity | 97% |
Physical Form | Powder |
Chemical Name or Material | 4-Boc-1-(5-bromo-2-pyridyl)piperazine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'