Learn More
4-(Boc-amino)-1-methylpyrrole-2-carboxylic Acid, TRC
Product Code. 30346591
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30346591 1 g 1g
1 options
Product Code. 30346591 Supplier Toronto Research Chemicals Supplier No. TRCB6185851G
 Restricted Item
Beräknad leverans: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-(Boc-amino)-1-methylpyrrole-2-carboxylic Acid
CAS 77716-11-1
Molecular Formula C11 H16 N2 O4
Quantity 1 g
Synonym 4-tert-Butoxycarbonylamino-1-methyl-1H-pyrrole-2-carboxylic Acid, 4-[[(1,1-Dimethylethoxy)carbonyl]amino]-1-methyl-1H-pyrrole-2-carboxylic Acid
InChI Formula InChI=1S/C11H16N2O4/c1-11(2,3)17-10(16)12-7-5-8(9(14)15)13(4)6-7/h5-6H,1-4H3,(H,12,16)(H,14,15)
SMILES Cn1cc(NC(=O)OC(C)(C)C)cc1C(=O)O
IUPAC Name 1-methyl-4-[(2-methylpropan-2-yl)oxycarbonylamino]pyrrole-2-carboxylic acid
Molecular Weight (g/mol) 240.256
Formula Weight 240.111