missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Boc-amino)tetrahydropyran-4-carboxylic acid, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 mg
250 mg
1 g
Description
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as pharmaceutical intermediate.
Solubility
Soluble in water. (15 g/L) (25°C)
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Used as pharmaceutical intermediate.
Solubility
Soluble in water. (15 g/L) (25°C)
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 157°C (decomposition) |
Quantity | 50 mg |
Solubility Information | Soluble in water. (15g/L) (25°C) |
Formula Weight | 245.27 |
Percent Purity | 95% |
Chemical Name or Material | 4-(Boc-amino)tetrahydropyran-4-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'