Läs mer
4-(Boc-aminomethyl)benzoic Acid, TRC
Produktkod. 30344187
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30344187 1 g
1 options
Produktkod. 30344187 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6192651G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-(Boc-aminomethyl)benzoic Acid
CAS 33233-67-9
Molekylformel C13H17NO4
Kvantitet 1 g
Synonym 4-[[[(1,1-Dimethylethoxy)carbonyl]amino]methyl]-benzoic Acid, α-(Carboxyamino)-p-toluic Acid N-tert-Butyl Ester, 4-(N-tert-Butoxycarbonylaminomethyl)benzoic Acid, 4-(t-Butoxycarbonylamino-methyl)benzoic Acid, 4-[(Boc-amino)methyl]benzoic Acid, 4-[[(tert-Butoxycarbonyl)amino]methyl]benzoic Acid, 4-[[[[(1,1-Dimethylethyl)oxy]carbonyl]amino]methyl]benzoic Acid, N-tert-Butoxycarbonyl-4-aminomethylbenzoic Acid
InChI formel InChI=1S/C13H17NO4/c1-13(2,3)18-12(17)14-8-9-4-6-10(7-5-9)11(15)16/h4-7H,8H2,1-3H3,(H,14,17)(H,15,16)
LEDER O=C(O)C1=CC=C(CNC(OC(C)(C)C)=O)C=C1
IUPAC-namn 4-(((tert-butoxycarbonyl)amino)methyl)benzoic acid
Molekylvikt (g/mol) 251.28
Formel vikt 251.12