Läs mer
4-Bromo-1-chloro-2-(4-ethoxybenzyl)benzene, TRC
Produktkod. 30138569
Change view
Kvantitet:
100 mg
1 g
2.5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30138569 1 g -
30011883 2.5 g 2.5g
17924595 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138569 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6821751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 461432-23-5
Molekylformel C15 H14 Br Cl O
Molekylvikt (g/mol) 325.63
Synonym 4-bromo-1-chloro-2-[(4-ethoxyphenyl)methyl]benzene, 4-Bromo-1-chloro-2-(4-ethoxybenzyl)benzene, 5-bromo-2-chloro-4'-ethoxydiphenylmethane
IUPAC-namn 4-bromo-1-chloro-2-[(4-ethoxyphenyl)methyl]benzene
LEDER CCOc1ccc(Cc2cc(Br)ccc2Cl)cc1
Specifikationer
Kemiskt namn eller material 4-Bromo-1-chloro-2-(4-ethoxybenzyl)benzene
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C15H14BrClO/c1-2-18-14-6-3-11(4-7-14)9-12-10-13(16)5-8-15(12)17/h3-8,10H,2,9H2,1H3
Formel vikt 323.9917
Procent renhet >95%