missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole, 97%, Thermo Scientific™

Catalog Number p-3075605
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
497832-99-2
C5H4BrF3N2
229
MFCD04115415
TZAYNGPUOOUEAP-UHFFFAOYSA-N
4-bromo-1-methyl-3-trifluoromethyl-1h-pyrazole, 4-bromo-1-methyl-3-trifluoromethyl pyrazole, 1h-pyrazole,4-bromo-1-methyl-3-trifluoromethyl, 1h-pyrazole, 4-bromo-1-methyl-3-trifluoromethyl, pubchem16983, buttpark 99\18-60, 4-bromo-1-methyl-3-trifluoromethyl pyrazol, 4-bromo-1-methyl-3-trifluoromethyl ;-1h-pyrazole
2794651
4-bromo-1-methyl-3-(trifluoromethyl)pyrazole
CN1C=C(C(=N1)C(F)(F)F)Br
This item is not returnable. View return policy
497832-99-2
C5H4BrF3N2
229
MFCD04115415
TZAYNGPUOOUEAP-UHFFFAOYSA-N
4-bromo-1-methyl-3-trifluoromethyl-1h-pyrazole, 4-bromo-1-methyl-3-trifluoromethyl pyrazole, 1h-pyrazole,4-bromo-1-methyl-3-trifluoromethyl, 1h-pyrazole, 4-bromo-1-methyl-3-trifluoromethyl, pubchem16983, buttpark 99\18-60, 4-bromo-1-methyl-3-trifluoromethyl pyrazol, 4-bromo-1-methyl-3-trifluoromethyl ;-1h-pyrazole
2794651
4-bromo-1-methyl-3-(trifluoromethyl)pyrazole
CN1C=C(C(=N1)C(F)(F)F)Br
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 229
Percent Purity 97%
Chemical Name or Material 4-Bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'