Learn More
4-Bromo-2-fluoro-1-iodobenzene, 98%, Thermo Scientific Chemicals
Catalog Number p-7022905
Click to view available options
Quantity:
25 g
100 g
105931-73-5
C6H3BrFI
300.90
MFCD00010608
XRMZKCQCINEBEI-UHFFFAOYSA-N
1-bromo-3-fluoro-4-iodobenzene, 3-fluoro-4-iodobromobenzene, 2-fluoro-4-bromoiodobenzene, 4-bromo-2-fluoroiodobenzene, 4-bromo-2-fluoro-1-iodo-benzene, benzene, 4-bromo-2-fluoro-1-iodo, pubchem3232, acmc-2098hz, 2-fluoro-4-bromo iodobenzene, 3-fluoro-4-iodo-bromobenzene
2724516
4-bromo-2-fluoro-1-iodobenzene
FC1=C(I)C=CC(Br)=C1
105931-73-5
C6H3BrFI
300.90
MFCD00010608
XRMZKCQCINEBEI-UHFFFAOYSA-N
1-bromo-3-fluoro-4-iodobenzene, 3-fluoro-4-iodobromobenzene, 2-fluoro-4-bromoiodobenzene, 4-bromo-2-fluoroiodobenzene, 4-bromo-2-fluoro-1-iodo-benzene, benzene, 4-bromo-2-fluoro-1-iodo, pubchem3232, acmc-2098hz, 2-fluoro-4-bromo iodobenzene, 3-fluoro-4-iodo-bromobenzene
2724516
4-bromo-2-fluoro-1-iodobenzene
FC1=C(I)C=CC(Br)=C1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 46°C to 50°C
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 4740250
Sensitivity Light sensitive
Formula Weight 300.9
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-fluoro-1-iodobenzene