missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-2-fluoro-N-(2-fluoro-3-methylphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64714.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449096

  • 2603.20 SEK / 1g


Estimated Shipment date
from Supplier 14-11-2018
Add to basket

Specifications

Chemical Identifiers

MFCD27975694

Specifications

4-Bromo-2-fluoro-N-(2-fluoro-3-methylphenyl)benzenesulfonamide
MFCD27975694
Solid
1g
362.19
97%
Videos
SDS