Learn More
Alfa Aesar™ 4-Bromo-2-fluoro-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H64940.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15479266
  • 2362.17 SEK / 1g
Estimated Shipment date
from Supplier 30-01-2018
Add to basket
Specifications
Chemical Identifiers
4-Bromo-2-fluoro-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
398.17
MFCD27975702
Specifications
4-Bromo-2-fluoro-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
97%
398.17
Solid
1g
MFCD27975702