Learn More
Alfa Aesar™ 4-Bromo-2-fluoro-N-isobutylbenzenesulfonamide, 97%
Brand:  Alfa Aesar™ H65991.MD
Additional Details : Weight : 0.00025kg
Quantity 250mg
 View more versions of this product
Product Code. 15470077
  • 809.89 SEK / 250mg
Estimated Shipment date
from Supplier 06-07-2018
Add to basket
Specifications
Chemical Identifiers
4-Bromo-2-fluoro-N-isobutylbenzenesulfonamide
310.18
MFCD27392033
20408826
Specifications
4-Bromo-2-fluoro-N-isobutylbenzenesulfonamide
Solid
250mg
MFCD27392033
149° to 150°C
97%
310.18
20408826