missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-2-methoxybenzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Bromo-2-methoxybenzaldehyde is used in the building block, synthesis of a nonacid degradable linker for solid-phase synthesis.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
4-Bromo-2-methoxybenzaldehyde is used in the building block, synthesis of a nonacid degradable linker for solid-phase synthesis.
Solubility
Soluble in water.
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 67°C to 71°C |
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Synonym | 4-bromo-2-methoxy-benzaldehyde, benzaldehyde, 4-bromo-2-methoxy, 2-methoxy-4-bromobenzaldehyde, 4-bromo-2-methoxy benzaldehyde, acmc-209jte, 4-bromo-2-methoxybezaldehyde, ksc494s3l, 4-bromo-2-methoxybenz-aldehyde, benzaldehyde,4-bromo-2-methoxy, 4-bromanyl-2-methoxy-benzaldehyde |
Solubility Information | Soluble in water. |
Formula Weight | 215.05 |
Percent Purity | 97% |
Chemical Name or Material | 4-Bromo-2-methoxybenzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'