missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxybenzyl alcohol, 97%, Thermo Scientific™

Catalog Number p-7029827
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
17102-63-5
C8H9BrO2
217.06
MFCD07369762
HBEIHPSICGGZIF-UHFFFAOYSA-N
15128243
(4-bromo-2-methoxyphenyl)methanol
COC1=C(CO)C=CC(Br)=C1
This item is not returnable. View return policy
17102-63-5
C8H9BrO2
217.06
MFCD07369762
HBEIHPSICGGZIF-UHFFFAOYSA-N
15128243
(4-bromo-2-methoxyphenyl)methanol
COC1=C(CO)C=CC(Br)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in azaquinone-methide-mediated depolymerization of aromatic carbamate oligomers.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 66°C to 70°C
Quantity 1 g
Synonym 4-bromo-2-methoxybenzyl alcohol, 4-bromo-2-methoxyphenyl methanol, 4-bromo-2-methoxy-phenyl-methanol, benzenemethanol, 4-bromo-2-methoxy, acmc-209e2a, 2-methoxy-4-bromobenzyl alcohol, benzenemethanol,4-bromo-2-methoxy
Solubility Information Insoluble in water.
Formula Weight 217.06
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methoxybenzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'