missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-methoxypyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-7077335
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
100367-39-3
C6H6BrNO
188.02
YFTGMMXMLPTTAY-UHFFFAOYSA-N
2-methoxy-4-bromopyridine, 4-bromo-2-methoxy-pyridine, pyridine, 4-bromo-2-methoxy, pubchem6657, ksc494g1d, acmc-1c790, abbypharma ap-17-5965
14062309
4-bromo-2-methoxypyridine
COC1=NC=CC(=C1)Br
This item is not returnable. View return policy
100367-39-3
C6H6BrNO
188.02
YFTGMMXMLPTTAY-UHFFFAOYSA-N
2-methoxy-4-bromopyridine, 4-bromo-2-methoxy-pyridine, pyridine, 4-bromo-2-methoxy, pubchem6657, ksc494g1d, acmc-1c790, abbypharma ap-17-5965
14062309
4-bromo-2-methoxypyridine
COC1=NC=CC(=C1)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 87.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 188.02
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methoxypyridine
compliance-icons
Product Identifier
  • 4-Bromo-2-methoxypyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'