missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-nitrophenylhydrazine hydrochloride, 95%, Thermo Scientific Chemicals

Catalog Number p-7030500
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
100032-79-9
C6H7BrClN3O2
268.50
MFCD09260482
SIBURLORIWZQNT-UHFFFAOYSA-N
4-bromo-2-nitrophenylhydrazine hydrochloride, 4-bromo-2-nitrophenyl hydrazine hydrochloride, acmc-20amnd, 4-bromo-2-nitrophenyl hydrazine;hydrochloride, 4-bromo-2-nitrophenylhydrazinehydrochloride, 4-bromo-2-nitrophenyl hydrazine-hydrogen chloride 1/1
44119379
(4-bromo-2-nitrophenyl)hydrazine;hydrochloride
Cl.NNC1=C(C=C(Br)C=C1)[N+]([O-])=O
This item is not returnable. View return policy
100032-79-9
C6H7BrClN3O2
268.50
MFCD09260482
SIBURLORIWZQNT-UHFFFAOYSA-N
4-bromo-2-nitrophenylhydrazine hydrochloride, 4-bromo-2-nitrophenyl hydrazine hydrochloride, acmc-20amnd, 4-bromo-2-nitrophenyl hydrazine;hydrochloride, 4-bromo-2-nitrophenylhydrazinehydrochloride, 4-bromo-2-nitrophenyl hydrazine-hydrogen chloride 1/1
44119379
(4-bromo-2-nitrophenyl)hydrazine;hydrochloride
Cl.NNC1=C(C=C(Br)C=C1)[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∽185°C (decomposition)
Quantity 1 g
Formula Weight 268.5
Percent Purity 95%
Chemical Name or Material 4-Bromo-2-nitrophenylhydrazine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'