Läs mer
4-Bromo-2-(trifluoromethyl)benzonitrile, TRC
Produktkod. 30343439
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30343439 100 mg
30331856 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30343439 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB695508100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Bromo-2-(trifluoromethyl)benzonitrile
CAS 191165-13-6
Molekylformel C8H3BrF3N
Kvantitet 100 mg
InChI formel InChI=1S/C8H3BrF3N/c9-6-2-1-5(4-13)7(3-6)8(10,11)12/h1-3H
LEDER FC(F)(F)c1cc(Br)ccc1C#N
IUPAC-namn 4-bromo-2-(trifluoromethyl)benzonitrile
Molekylvikt (g/mol) 250.02
Formel vikt 248.9401