Läs mer
(4-Bromo-3-fluorophenyl)carbamic Acid Benzyl Ester, TRC
Produktkod. 30338726
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30338726 100 mg
1 options
Produktkod. 30338726 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB683310100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (4-Bromo-3-fluorophenyl)carbamic Acid Benzyl Ester
CAS 510729-01-8
Molekylformel C14H11BrFNO2
Kvantitet 100 mg
Synonym (4-Bromo-3-fluorophenyl)carbamic Acid Phenylmethyl Ester, N-(4-Bromo-3-fluorophenyl)carbamic Acid Phenylmethyl Ester
InChI formel InChI=1S/C14H11BrFNO2/c15-12-7-6-11(8-13(12)16)17-14(18)19-9-10-4-2-1-3-5-10/h1-8H,9H2,(H,17,18)
LEDER BrC1=CC=C(NC(OCC2=CC=CC=C2)=O)C=C1F
IUPAC-namn benzyl N-(4-bromo-3-fluorophenyl)carbamate
Molekylvikt (g/mol) 324.15
Formel vikt 323