missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-3-(trifluoromethyl)benzonitrile, 98%

Brand:  Alfa Aesar™ H61459.06

Additional Details : CAS Number : 1735-53-1 Weight : 0.00500kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15458008

  • 3181.69 SEK / 5g


Estimated Shipment date
from Supplier 13-08-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-3-(trifluoromethyl)benzonitrile
1735-53-1
MFCD03095347
UN3439

Specifications

4-Bromo-3-(trifluoromethyl)benzonitrile
1735-53-1
98%
Pale yellow
UN3439
KSXUIQQDHHFSRN-UHFFFAOYSA-N
250.018
250.02
Crystalline Solid
C8H3BrF3N
MFCD03095347
5g
C1=CC(=C(C=C1C#N)C(F)(F)F)Br
10800647
Videos
SDS