Läs mer
4-Bromo-Alpha-phenylbenzenemethanol, TRC
Produktkod. 30346511
Change view
Kvantitet:
2.5 g
Produktkod. Kvantitet
30346511 2.5 g
1 options
Produktkod. 30346511 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6802702.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Bromo-alpha-phenylbenzenemethanol
CAS 29334-16-5
Molekylformel C13 H11 Br O
Kvantitet 2.5 g
Synonym 4-Bromo-α-phenylbenzenemethanol, Benzhydrol, 4-bromo- (8CI), Benzohydrol, 4-bromo- (4CI), 4-Bromo-α-phenylbenzenemethanol, (4-Bromophenyl)phenylmethanol, (p-Bromophenyl)-α-phenylmethanol, (±)-(4-Bromophenyl)phenylmethanol, 4-(1-Hydroxybenzyl)bromobenzene, 4-Bromobenzhydrol, NSC 89817, p-Bromobenzhydrol, α-(4-Bromophenyl)benzenemethanol, α-Phenyl-p-bromobenzyl alcohol
InChI formel InChI=1S/C13H11BrO/c14-12-8-6-11(7-9-12)13(15)10-4-2-1-3-5-10/h1-9,13,15H
LEDER OC(c1ccccc1)c2ccc(Br)cc2
IUPAC-namn (4-bromophenyl)-phenylmethanol
Molekylvikt (g/mol) 263.13
Formel vikt 261.9993