missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-m-xylene, 97%, Thermo Scientific™

Catalog Number p-75755
Click to view available options
Quantity:
10 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
583-70-0
C8H9Br
185.06
MFCD00000071
YSFLQVNTBBUKEA-UHFFFAOYSA-N
4-bromo-m-xylene, 2,4-dimethylbromobenzene, benzene, 1-bromo-2,4-dimethyl, 4-bromo-1,3-dimethylbenzene, 1,3-dimethyl-4-bromobenzene, m-xylene, 4-bromo, 1-bromo-2,4-dimethyl-benzene, 4-brom-m-xylol, pubchem3762, acmc-1aowv
68503
1-bromo-2,4-dimethylbenzene
CC1=CC=C(Br)C(C)=C1
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
583-70-0
C8H9Br
185.06
MFCD00000071
YSFLQVNTBBUKEA-UHFFFAOYSA-N
4-bromo-m-xylene, 2,4-dimethylbromobenzene, benzene, 1-bromo-2,4-dimethyl, 4-bromo-1,3-dimethylbenzene, 1,3-dimethyl-4-bromobenzene, m-xylene, 4-bromo, 1-bromo-2,4-dimethyl-benzene, 4-brom-m-xylol, pubchem3762, acmc-1aowv
68503
1-bromo-2,4-dimethylbenzene
CC1=CC=C(Br)C(C)=C1

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
Color Colorless
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula (CH3)2C6H3Br
Beilstein 05, II, 293
Formula Weight 185.06
Percent Purity ≥96.0%
Quantity 10 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5500 to 1.5520 (20°C, 589nm)
Chemical Name or Material 4-Bromo-m-xylene, 97%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'