missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2,3-dichlorophenyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64261.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15448806

  • 2603.20 SEK / 1g


Estimated Shipment date
from Supplier 12-11-2018
Add to basket

Specifications

Chemical Identifiers

MFCD28098717

Specifications

4-Bromo-N-(2,3-dichlorophenyl)-2-fluorobenzenesulfonamide
MFCD28098717
Solid
1g
399.06
97%
Videos
SDS