Learn More
Alfa Aesar™ 4-Bromo-N-(2,5-difluorophenyl)-2-fluorobenzenesulfonamide, 97%
Brand:  Alfa Aesar™ H64694.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15449076
  • 2603.20 SEK / 1g
Estimated Shipment date
from Supplier 16-11-2018
Add to basket
Specifications
Chemical Identifiers
MFCD27975692
Specifications
4-Bromo-N-(2,5-difluorophenyl)-2-fluorobenzenesulfonamide
MFCD27975692
Solid
1g
366.15
97%