missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2,5-difluorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63135.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498136

  • 3427.64 SEK / 1g


Estimated Shipment date
from Supplier 16-07-2019
Add to basket

Specifications

Chemical Identifiers

MFCD26143354

Specifications

4-Bromo-N-(2,5-difluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
MFCD26143354
Solid
1g
416.16
97%
Videos
SDS