missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2,5-dimethylphenyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64454.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498926

  • 2716.64 SEK / 1g


Estimated Shipment date
from Supplier 18-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD21364183

Specifications

4-Bromo-N-(2,5-dimethylphenyl)-2-fluorobenzenesulfonamide
MFCD21364183
Solid
1g
358.23
97%
Videos
SDS