missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-bromo-4-fluorophenyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64778.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489136

  • 2603.20 SEK / 1g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-bromo-4-fluorophenyl)-2-fluorobenzenesulfonamide
427.06
MFCD27975698

Specifications

4-Bromo-N-(2-bromo-4-fluorophenyl)-2-fluorobenzenesulfonamide
1g
427.06
Solid
97%
MFCD27975698
Videos
SDS