missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-chloro-4-methylphenyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64614.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489016

  • 2362.17 SEK / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-chloro-4-methylphenyl)-2-fluorobenzenesulfonamide
378.64
MFCD28098727

Specifications

4-Bromo-N-(2-chloro-4-methylphenyl)-2-fluorobenzenesulfonamide
1g
378.64
Solid
97%
MFCD28098727
Videos
SDS