missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-ethoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63243.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15478186

  • 3114.20 SEK / 1g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-ethoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
424.23
MFCD26143356

Specifications

4-Bromo-N-(2-ethoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
1g
424.23
Solid
97%
MFCD26143356
Videos
SDS