Learn More
Alfa Aesar™ 4-Bromo-N-(2-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H62253.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15487206
  • 2834.60 SEK / 1g
Estimated Shipment date
from Supplier 05-03-2018
Add to basket
Specifications
Chemical Identifiers
4-Bromo-N-(2-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
398.17
MFCD26143318
Specifications
4-Bromo-N-(2-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
97%
398.17
Solid
1g
MFCD26143318