Learn More
Alfa Aesar™ 4-Bromo-N-(2-isopropylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H63556.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15468366
  • 3114.20 SEK / 1g
Estimated Shipment date
from Supplier 07-05-2018
Add to basket
Specifications
Chemical Identifiers
4-Bromo-N-(2-isopropylphenyl)-3-(trifluoromethyl)benzenesulfonamide
422.26
MFCD26143374
Specifications
4-Bromo-N-(2-isopropylphenyl)-3-(trifluoromethyl)benzenesulfonamide
1g
422.26
Solid
97%
MFCD26143374