missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(2-methoxybenzyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63080.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15458116

  • 11319.11 SEK / 5g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(2-methoxybenzyl)-3-(trifluoromethyl)benzenesulfonamide
424.23
MFCD26960687

Specifications

4-Bromo-N-(2-methoxybenzyl)-3-(trifluoromethyl)benzenesulfonamide
5g
424.23
Solid
97%
MFCD26960687
Videos
SDS