Learn More
Alfa Aesar™ 4-Bromo-N-(2-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H63944.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15488586
  • 2834.60 SEK / 1g
Estimated Shipment date
from Supplier 05-02-2018
Add to basket
Specifications
Chemical Identifiers
4-Bromo-N-(2-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
410.21
MFCD26960690
Specifications
4-Bromo-N-(2-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
1g
410.21
Solid
97%
MFCD26960690