Learn More
Alfa Aesar™ 4-Bromo-N-(2-methylbenzyl)-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H62372.03
UNSPSC : 12352105
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15487336
  • 3035.71 SEK / 1g
Estimated Shipment date
from Supplier 04-09-2017
Add to basket
Specifications
Chemical Identifiers
4-Bromo-N-(2-methylbenzyl)-3-(trifluoromethyl)benzenesulfonamide
408.23
MFCD26143324
Specifications
4-Bromo-N-(2-methylbenzyl)-3-(trifluoromethyl)benzenesulfonamide
1g
408.23
Solid
97%
MFCD26143324
Documentation
Technical Documents