Learn More
Alfa Aesar™ 4-Bromo-N-(3,5-difluorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H63618.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15458406
  • 2834.60 SEK / 1g
Estimated Shipment date
from Supplier 06-02-2018
Add to basket
Specifications
Chemical Identifiers
4-Bromo-N-(3,5-difluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
416.16
MFCD26143377
Specifications
4-Bromo-N-(3,5-difluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
1g
416.16
Solid
97%
MFCD26143377