Learn More
Alfa Aesar™ 4-Bromo-N-(3-fluoro-5-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H63570.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15448376
  • 2834.60 SEK / 1g
Estimated Shipment date
from Supplier 07-03-2018
Add to basket
Specifications
Chemical Identifiers
4-Bromo-N-(3-fluoro-5-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
412.2
MFCD26143376
Specifications
4-Bromo-N-(3-fluoro-5-methylphenyl)-3-(trifluoromethyl)benzenesulfonamide
97%
412.2
Solid
1g
MFCD26143376