missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(3-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H62751.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15467806

  • 12447.17 SEK / 5g


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(3-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
398.17
MFCD26143336

Specifications

4-Bromo-N-(3-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
97%
398.17
Solid
5g
MFCD26143336
Videos
SDS