missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(3-isopropylphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63715.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15488466

  • 3114.20 SEK / 1g


Estimated Shipment date
from Supplier 02-08-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(3-isopropylphenyl)-3-(trifluoromethyl)benzenesulfonamide
MFCD27665233

Specifications

4-Bromo-N-(3-isopropylphenyl)-3-(trifluoromethyl)benzenesulfonamide
Solid
97%
422.26
1g
MFCD27665233
Videos
SDS