missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-bromo-3-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H63068.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15498106

  • 3427.64 SEK / 1g


Estimated Shipment date
from Supplier 24-07-2019
Add to basket

Specifications

Chemical Identifiers

MFCD26143353

Specifications

4-Bromo-N-(4-bromo-3-fluorophenyl)-3-(trifluoromethyl)benzenesulfonamide
MFCD26143353
Solid
1g
477.07
97%
Videos
SDS