Learn More
Alfa Aesar™ 4-Bromo-N-(4-bromo-3-methylphenyl)-2-fluorobenzenesulfonamide, 97%
Brand:  Alfa Aesar™ H64656.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15449056
  • 2603.20 SEK / 1g
Estimated Shipment date
from Supplier 12-06-2018
Add to basket
Specifications
Chemical Identifiers
4-Bromo-N-(4-bromo-3-methylphenyl)-2-fluorobenzenesulfonamide
423.1
MFCD27975689
Specifications
4-Bromo-N-(4-bromo-3-methylphenyl)-2-fluorobenzenesulfonamide
1g
423.1
Solid
97%
MFCD27975689