missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Bromo-N-(4-bromophenyl)-2-fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64767.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449136

  • 2362.17 SEK / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

4-Bromo-N-(4-bromophenyl)-2-fluorobenzenesulfonamide
409.07
MFCD27824752

Specifications

4-Bromo-N-(4-bromophenyl)-2-fluorobenzenesulfonamide
Solid
409.07
1g
97%
MFCD27824752
Videos
SDS