Learn More
Alfa Aesar™ 4-Bromo-N-(4-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H63654.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15448426
  • 3427.64 SEK / 1g
Estimated Shipment date
from Supplier 14-12-2018
Add to basket
Specifications
Chemical Identifiers
MFCD26143379
Specifications
4-Bromo-N-(4-methoxyphenyl)-3-(trifluoromethyl)benzenesulfonamide
MFCD26143379
Solid
1g
410.21
97%